Product Details

SNP ID
rs117562382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:41484481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATGGACTAGATTGGCCCCCTGA[C/T]CATAGGGGTTTCCATCAATCATGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KC6 PubMed Links

Gene Details

Gene
KC6
Gene Name
keratoconus gene 6
There are no transcripts associated with this gene.

View Full Product Details