Product Details

SNP ID
rs114043670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:49563442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTCATTGCAAAAAGTATTTACT[A/G]CATACCTACTTAGTGTCAGACACTA
Phenotype
MIM: 603684
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIPG PubMed Links

Gene Details

Gene
LIPG
Gene Name
lipase G, endothelial type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308006.1 Intron NP_001294935.1
NM_006033.3 Intron NP_006024.1
XM_005258390.1 Intron XP_005258447.1
XM_011526265.2 Intron XP_011524567.1
XM_011526267.1 Intron XP_011524569.1
XM_017026095.1 Intron XP_016881584.1

View Full Product Details