Product Details

SNP ID
rs116813540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41387131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGAGACTCCCAGTCTTCACTGC[C/T]ATGTATTCCTAGATTCATGAGGTCC
Phenotype
MIM: 606492
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC5 PubMed Links

Gene Details

Gene
EXOSC5
Gene Name
exosome component 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020158.3 Intron NP_064543.3
Gene
TMEM91
Gene Name
transmembrane protein 91
There are no transcripts associated with this gene.

View Full Product Details