Product Details

SNP ID
hCV153277807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41715103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGGTAAACAATCAGAGCCTCC[A/C]GGTCAGTCCCAGGCTGCAGCTGTCC
Phenotype
MIM: 114890
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CEACAM5 PubMed Links

Gene Details

Gene
CEACAM5
Gene Name
carcinoembryonic antigen related cell adhesion molecule 5
There are no transcripts associated with this gene.

View Full Product Details