Product Details

SNP ID
rs117813514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42223173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTGCACGCAGGCAAAGGCTATG[A/G]TATTCTCAGAAAAGGTGTGCCTGGC
Phenotype
MIM: 606784 MIM: 614387
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEDD2 PubMed Links

Gene Details

Gene
DEDD2
Gene Name
death effector domain containing 2
There are no transcripts associated with this gene.

Gene
GSK3A
Gene Name
glycogen synthase kinase 3 alpha
There are no transcripts associated with this gene.

Gene
ZNF526
Gene Name
zinc finger protein 526
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314033.1 Intron NP_001300962.1
NM_133444.2 Intron NP_597701.1

View Full Product Details