Product Details

SNP ID
rs115237503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44025447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCGCAACCACCCAATGATCGATT[C/T]AGGAGAAAAGAAGCCTGGGCGGAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF222 PubMed Links

Gene Details

Gene
ZNF222
Gene Name
zinc finger protein 222
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129996.1 124 Missense Mutation TCA,TTA S4L NP_001123468.1
NM_013360.2 124 UTR 5 NP_037492.2
XM_005259208.3 124 UTR 5 XP_005259265.1
XM_005259209.3 124 Missense Mutation TCA,TTA S4L XP_005259266.1
XM_017027237.1 124 Missense Mutation TCA,TTA S4L XP_016882726.1
XM_017027238.1 124 UTR 5 XP_016882727.1

View Full Product Details