Product Details

SNP ID
rs115919347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48661687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCCATCTCACGTCTAGTGCTCC[A/G]GCCTGGGACTGGGTGTGGGTGCCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NTN5 PubMed Links

Gene Details

Gene
NTN5
Gene Name
netrin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145807.2 1630 Missense Mutation CCG,CTG P487L NP_665806.1
XM_011526443.2 1630 Intron XP_011524745.1
XM_017026274.1 1630 Missense Mutation CCG,CTG P332L XP_016881763.1
Gene
SEC1P
Gene Name
secretory blood group 1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details