Product Details

SNP ID
rs114601492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55159283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGGGGCGGAGTTCCGGGTTCCA[C/T]AGCTGGGACAGTGTTGCCCAGAGCT
Phenotype
MIM: 614566 MIM: 191044 MIM: 191041
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAAF3 PubMed Links

Gene Details

Gene
DNAAF3
Gene Name
dynein axonemal assembly factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256714.1 1620 Missense Mutation ATG,GTG M536V NP_001243643.1
NM_001256715.1 1620 Missense Mutation ATG,GTG M469V NP_001243644.1
NM_001256716.1 1620 Missense Mutation ATG,GTG M415V NP_001243645.1
NM_178837.4 1620 Missense Mutation ATG,GTG M516V NP_849159.2
Gene
TNNI3
Gene Name
troponin I3, cardiac type
There are no transcripts associated with this gene.

Gene
TNNT1
Gene Name
troponin T1, slow skeletal type
There are no transcripts associated with this gene.

View Full Product Details