Product Details
- SNP ID
-
rs116236774
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:55432452 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCAAGGCACCTTCTCTGGTGATG[A/G]CATCACAGGAAGGAGGCAGGGTCCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SHISA7
PubMed Links
Gene Details
- Gene
- SHISA7
- Gene Name
- shisa family member 7
View Full Product Details