Product Details
- SNP ID
-
rs114577962
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:56087607 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTCTAAAAGAGAAAAGGGCCCCT[A/G]GTTCTCTTCCCTCTCTGGGATCCTC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101928886
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150405930] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101928886
- Gene Name
- uncharacterized LOC101928886
There are no transcripts associated with this gene.
- Gene
- ZNF787
- Gene Name
- zinc finger protein 787
View Full Product Details