Product Details

SNP ID
rs111393043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57686519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGAGAATGAGGCGATAGCTTCT[A/G]AGCAGAGTGTATCTATACAGGTCAG
Phenotype
MIM: 604085 MIM: 613419
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF154 PubMed Links

Gene Details

Gene
ZNF154
Gene Name
zinc finger protein 154
There are no transcripts associated with this gene.

Gene
ZNF551
Gene Name
zinc finger protein 551
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270938.1 439 Intron NP_001257867.1
NM_138347.4 439 Missense Mutation AAG,GAG K82E NP_612356.2
Gene
ZSCAN4
Gene Name
zinc finger and SCAN domain containing 4
There are no transcripts associated with this gene.

View Full Product Details