Product Details

SNP ID
rs116889879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58476564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCTGTGGCCATCCCTCGACTCC[A/G]CGCAACGTGGAGCTTTGGCCCCGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF324 PubMed Links

Gene Details

Gene
ZNF324
Gene Name
zinc finger protein 324
There are no transcripts associated with this gene.

Gene
ZNF446
Gene Name
zinc finger protein 446
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304453.1 Intron NP_001291382.1
NM_017908.3 Intron NP_060378.1
XM_005259052.4 Intron XP_005259109.1
XM_005259053.4 Intron XP_005259110.1
XM_006723266.3 Intron XP_006723329.1
XM_017026949.1 Intron XP_016882438.1
XM_017026950.1 Intron XP_016882439.1

View Full Product Details