Product Details
- SNP ID
-
rs113551985
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:35890025 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCCGGCCCAGGGGGCAGGGCAGC[C/T]GCCATGTGGAGGGCTGTGTTCCCGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NFKBID
PubMed Links
Gene Details
- Gene
- NFKBID
- Gene Name
- NFKB inhibitor delta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001321831.1 |
1071 |
Silent Mutation |
GCA,GCG |
A266A |
NP_001308760.1 |
NM_139239.2 |
1071 |
Silent Mutation |
GCA,GCG |
A251A |
NP_640332.1 |
XM_005259340.3 |
1071 |
Silent Mutation |
GCA,GCG |
A266A |
XP_005259397.1 |
XM_011527419.2 |
1071 |
Silent Mutation |
GCA,GCG |
A403A |
XP_011525721.1 |
XM_011527420.2 |
1071 |
Silent Mutation |
GCA,GCG |
A266A |
XP_011525722.1 |
XM_011527421.2 |
1071 |
Silent Mutation |
GCA,GCG |
A241A |
XP_011525723.1 |
XM_011527424.2 |
1071 |
Silent Mutation |
GCA,GCG |
A266A |
XP_011525726.1 |
XM_017027389.1 |
1071 |
Silent Mutation |
GCA,GCG |
A266A |
XP_016882878.1 |
XM_017027390.1 |
1071 |
Silent Mutation |
GCA,GCG |
A266A |
XP_016882879.1 |
XM_017027391.1 |
1071 |
Silent Mutation |
GCA,GCG |
A251A |
XP_016882880.1 |
XM_017027392.1 |
1071 |
Silent Mutation |
GCA,GCG |
A251A |
XP_016882881.1 |
XM_017027393.1 |
1071 |
Silent Mutation |
GCA,GCG |
A251A |
XP_016882882.1 |
XM_017027394.1 |
1071 |
Silent Mutation |
GCA,GCG |
A241A |
XP_016882883.1 |
XM_017027395.1 |
1071 |
Silent Mutation |
GCA,GCG |
A241A |
XP_016882884.1 |
XM_017027396.1 |
1071 |
Intron |
|
|
XP_016882885.1 |
XM_017027397.1 |
1071 |
Silent Mutation |
GCA,GCG |
A218A |
XP_016882886.1 |
View Full Product Details