Product Details
- SNP ID
-
rs111599753
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:36639442 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCCTTCCCACATTCTTTACATTC[A/G]TAAGGTTTCTCACCAGTGTGAATTC
- Phenotype
-
MIM: 608640
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF461
PubMed Links
Gene Details
- Gene
- ZNF461
- Gene Name
- zinc finger protein 461
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001297623.2 |
1110 |
Silent Mutation |
TAC,TAT |
Y278Y |
NP_001284552.1 |
NM_001322821.1 |
1110 |
Silent Mutation |
TAC,TAT |
Y295Y |
NP_001309750.1 |
NM_001322823.1 |
1110 |
Silent Mutation |
TAC,TAT |
Y270Y |
NP_001309752.1 |
NM_001322825.1 |
1110 |
Silent Mutation |
TAC,TAT |
Y270Y |
NP_001309754.1 |
NM_001322826.1 |
1110 |
Silent Mutation |
TAC,TAT |
Y247Y |
NP_001309755.1 |
NM_001322827.1 |
1110 |
Silent Mutation |
TAC,TAT |
Y247Y |
NP_001309756.1 |
NM_001322828.1 |
1110 |
Silent Mutation |
TAC,TAT |
Y213Y |
NP_001309757.1 |
NM_153257.4 |
1110 |
Silent Mutation |
TAC,TAT |
Y301Y |
NP_694989.2 |
XM_011527486.2 |
1110 |
Silent Mutation |
TAC,TAT |
Y174Y |
XP_011525788.1 |
XM_017027470.1 |
1110 |
Silent Mutation |
TAC,TAT |
Y257Y |
XP_016882959.1 |
View Full Product Details