Product Details

SNP ID
rs117291365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3522215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCTTCTGTCTCCATGGCCTGCA[C/T]GGACCTTCACTTTATTCTTGTACAA
Phenotype
MIM: 603619
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FZR1 PubMed Links

Gene Details

Gene
FZR1
Gene Name
fizzy/cell division cycle 20 related 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136197.1 Intron NP_001129669.1
NM_001136198.1 Intron NP_001129670.1
NM_016263.3 Intron NP_057347.2
XM_005259573.4 Intron XP_005259630.1
XM_017026863.1 Intron XP_016882352.1

View Full Product Details