Product Details

SNP ID
rs116849418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3587220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAGAGACTGGGTAAAGTGGAGT[C/T]GGAGGAGGACGCGTCCAGAACCACT
Phenotype
MIM: 608792 MIM: 605535 MIM: 188070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIPC3 PubMed Links

Gene Details

Gene
GIPC3
Gene Name
GIPC PDZ domain containing family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133261.2 Intron NP_573568.1
XM_005259492.3 Intron XP_005259549.1
Gene
HMG20B
Gene Name
high mobility group 20B
There are no transcripts associated with this gene.

Gene
TBXA2R
Gene Name
thromboxane A2 receptor
There are no transcripts associated with this gene.

View Full Product Details