Product Details

SNP ID
rs116932400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:535893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTCCACCAGCCTTTCGGTTCCT[A/G]ACCAAGATGTGGCACCCTAACATCT
Phenotype
MIM: 116948 MIM: 600311
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC34 PubMed Links

Gene Details

Gene
CDC34
Gene Name
cell division cycle 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004359.1 433 Silent Mutation CTA,CTG L78L NP_004350.1
XM_005259690.3 433 Silent Mutation CTA,CTG L78L XP_005259747.1
XM_006722952.2 433 Silent Mutation CTA,CTG L78L XP_006723015.1
Gene
GZMM
Gene Name
granzyme M
There are no transcripts associated with this gene.

View Full Product Details