Product Details

SNP ID
rs114842563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10578865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCACCCACCCGAGATCATGCATT[A/G]TTAATAAGCATTCCCCCAAGGCCCA
Phenotype
MIM: 607309 MIM: 600927
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP1M2 PubMed Links

Gene Details

Gene
AP1M2
Gene Name
adaptor related protein complex 1 mu 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300887.1 Intron NP_001287816.1
NM_005498.4 Intron NP_005489.2
Gene
CDKN2D
Gene Name
cyclin dependent kinase inhibitor 2D
There are no transcripts associated with this gene.

View Full Product Details