Product Details

SNP ID
rs117998813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11832737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCCTATAAATGCGAGCAATGTG[A/G]GAAAGCCTTCAGAGCTGTGTCAATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF440 PubMed Links

Gene Details

Gene
ZNF440
Gene Name
zinc finger protein 440
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152357.2 1595 Missense Mutation AGA,GGA R521G NP_689570.2
XM_005259731.3 1595 Missense Mutation AGA,GGA R524G XP_005259788.1
XM_017026253.1 1595 Missense Mutation AGA,GGA R523G XP_016881742.1
XM_017026254.1 1595 Missense Mutation AGA,GGA R399G XP_016881743.1

View Full Product Details