Product Details

SNP ID
rs112833471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16067693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGAATGCCATCGACCGCGCGGA[A/G]CAGGCGGAGGCGGATAAGAAAGCCG
Phenotype
MIM: 600317
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TPM4 PubMed Links

Gene Details

Gene
TPM4
Gene Name
tropomyosin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145160.1 187 Silent Mutation GAA,GAG E23E NP_001138632.1
NM_003290.2 187 Intron NP_003281.1
XM_005260042.3 187 Intron XP_005260099.1
XM_006722865.2 187 Silent Mutation GAA,GAG E23E XP_006722928.1
XM_011528231.2 187 Intron XP_011526533.1
XM_011528232.1 187 Intron XP_011526534.1
XM_017027188.1 187 Intron XP_016882677.1
XM_017027189.1 187 Intron XP_016882678.1

View Full Product Details