Product Details

SNP ID
rs118003109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17938485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTGGTTTCCACACCCTGGGTA[A/G]CTGAGACACTGGCCCTCTAGGCAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC124 PubMed Links

Gene Details

Gene
CCDC124
Gene Name
coiled-coil domain containing 124
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136203.1 Intron NP_001129675.1
NM_138442.3 Intron NP_612451.1

View Full Product Details