Product Details

SNP ID
rs115645046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18564851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAGCTGGGCAGTGAAGCTCATG[C/T]CCTCTCTCCACCATCAGGCTGGACC
Phenotype
MIM: 615178 MIM: 191321
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KXD1 PubMed Links

Gene Details

Gene
KXD1
Gene Name
KxDL motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171948.1 Intron NP_001165419.1
NM_001171949.1 Intron NP_001165420.1
NM_024069.3 Intron NP_076974.2
XM_005260073.4 Intron XP_005260130.1
XM_006722883.2 Intron XP_006722946.1
XM_011528272.2 Intron XP_011526574.1
XM_011528273.2 Intron XP_011526575.1
XM_017027281.1 Intron XP_016882770.1
XM_017027282.1 Intron XP_016882771.1
Gene
UBA52
Gene Name
ubiquitin A-52 residue ribosomal protein fusion product 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033930.2 Intron NP_001029102.1
NM_001321017.1 Intron NP_001307946.1
NM_001321018.1 Intron NP_001307947.1
NM_001321019.1 Intron NP_001307948.1
NM_001321020.1 Intron NP_001307949.1
NM_001321021.1 Intron NP_001307950.1
NM_001321022.1 Intron NP_001307951.1
NM_003333.4 Intron NP_003324.1
XM_005260052.2 Intron XP_005260109.1
XM_005260053.3 Intron XP_005260110.1
XM_005260054.2 Intron XP_005260111.1
XM_006722871.2 Intron XP_006722934.1
XM_017027198.1 Intron XP_016882687.1

View Full Product Details