Product Details

SNP ID
rs112161622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63983216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAAATGACACCAATTACGATGAG[G/T]TGAGATGTGTCCGGCTTTCGTGGCT
Phenotype
MIM: 613979
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRPF6 PubMed Links

Gene Details

Gene
PRPF6
Gene Name
pre-mRNA processing factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012469.3 Intron NP_036601.2
XM_006723769.2 Intron XP_006723832.1
Gene
SAMD10
Gene Name
sterile alpha motif domain containing 10
There are no transcripts associated with this gene.

View Full Product Details