Product Details

SNP ID
rs115909300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:43559578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCTTGAAACAGGGACGATGAT[A/G]GTTCCTAACCCGTGGGGTTGTGGGA
Phenotype
MIM: 607589
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SGK2 PubMed Links

Gene Details

Gene
SGK2
Gene Name
SGK2, serine/threonine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199264.1 Intron NP_001186193.1
NM_016276.3 Intron NP_057360.2
NM_170693.2 Intron NP_733794.1

View Full Product Details