Product Details

SNP ID
rs113904719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45253152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCACTTCCCAGGCTTCCTCCTTG[C/T]TGGGTGAGAGTGAGGCTACCGGTCA
Phenotype
MIM: 107285
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLPI PubMed Links

Gene Details

Gene
SLPI
Gene Name
secretory leukocyte peptidase inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003064.3 Intron NP_003055.1

View Full Product Details