Product Details

SNP ID
rs115270794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46686897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGTTGGGATGGGTACTGAGGAA[C/G]GCCTTCTCCAGGACATAGAGGTCTA
Phenotype
MIM: 606411 MIM: 608679
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC13A3 PubMed Links

Gene Details

Gene
SLC13A3
Gene Name
solute carrier family 13 member 3
There are no transcripts associated with this gene.

Gene
TP53RK
Gene Name
TP53 regulating kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033550.3 841 Silent Mutation GCC,GCG A206A NP_291028.3

View Full Product Details