Product Details

SNP ID
rs114928867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49221483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTGCTGGGCAGACGACAAAAAG[C/T]TTTGGGGAAGAACCGCAGTGCTGAT
Phenotype
MIM: 616621 MIM: 601716
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX27 PubMed Links

Gene Details

Gene
DDX27
Gene Name
DEAD-box helicase 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017895.7 250 Missense Mutation GCT,GTT A73V NP_060365.7
XM_011528888.2 250 Missense Mutation GCT,GTT A73V XP_011527190.1
Gene
STAU1
Gene Name
staufen double-stranded RNA binding protein 1
There are no transcripts associated with this gene.

View Full Product Details