Product Details

SNP ID
rs115701570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50813891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGTCCTTCCTTGAATGTCACTT[A/G]AAGTGAGCCTCTCGCCTCCTTCCCC
Phenotype
MIM: 607471
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCAS4 PubMed Links

Gene Details

Gene
BCAS4
Gene Name
breast carcinoma amplified sequence 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010974.2 Intron NP_001010974.1
NM_017843.4 Intron NP_060313.3
NM_198799.3 Intron NP_942094.2
XM_011528886.2 Intron XP_011527188.1
XM_011528887.2 Intron XP_011527189.1
XM_017027932.1 Intron XP_016883421.1

View Full Product Details