Product Details

SNP ID
rs116754368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31465549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGACCGGTGGAGGTTTCAATGCA[C/T]AGGAGAGACAGCACAGGGACGCATC
Phenotype
MIM: 610388
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEFB124 PubMed Links

Gene Details

Gene
DEFB124
Gene Name
defensin beta 124
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037500.1 790 Missense Mutation TAT,TGT Y58C NP_001032589.1
XM_017027782.1 790 Missense Mutation TAT,TGT Y75C XP_016883271.1
Gene
REM1
Gene Name
RRAD and GEM like GTPase 1
There are no transcripts associated with this gene.

View Full Product Details