Product Details

SNP ID
rs117729108
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35003210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCAATGTAGCTAACGAGAGCA[C/G]AGGGTGACTGCCGGTCCGGGTTCAA
Phenotype
MIM: 609928 MIM: 608430
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
MYH7B PubMed Links
Additional Information
For this assay, SNP(s) [rs59909520] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MYH7B
Gene Name
myosin heavy chain 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020884.4 1842 Intron NP_065935.3
XM_006723840.3 1842 Intron XP_006723903.1
XM_011528941.2 1842 Intron XP_011527243.1
XM_011528947.2 1842 Intron XP_011527249.1
XM_011528948.2 1842 Intron XP_011527250.1
XM_017027986.1 1842 Intron XP_016883475.1
XM_017027987.1 1842 Intron XP_016883476.1
Gene
TRPC4AP
Gene Name
transient receptor potential cation channel subfamily C member 4 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015638.2 1842 Missense Mutation TCT,TGT S777C NP_056453.1
NM_199368.1 1842 Missense Mutation TCT,TGT S769C NP_955400.1
XM_011528772.2 1842 Intron XP_011527074.1
XM_011528774.1 1842 Missense Mutation TCT,TGT S379C XP_011527076.1
XM_017027799.1 1842 Intron XP_016883288.1
XM_017027800.1 1842 Missense Mutation TCT,TGT S439C XP_016883289.1
XM_017027801.1 1842 Missense Mutation TCT,TGT S379C XP_016883290.1

View Full Product Details