Product Details

SNP ID
rs112875918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18291961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGTTCCACCCCAGTGGTATATA[A/G]ACCATCCTTTCAAGAGTTTATCTTG
Phenotype
MIM: 604075
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF133 PubMed Links

Gene Details

Gene
ZNF133
Gene Name
zinc finger protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083330.2 2144 Intron NP_001076799.1
NM_001282995.1 2144 Intron NP_001269924.1
NM_001282996.1 2144 Intron NP_001269925.1
NM_001282997.1 2144 Intron NP_001269926.1
NM_001282998.1 2144 Intron NP_001269927.1
NM_001282999.1 2144 Intron NP_001269928.1
NM_001283000.1 2144 Intron NP_001269929.1
NM_001283001.1 2144 Intron NP_001269930.1
NM_001283002.1 2144 Intron NP_001269931.1
NM_001283003.1 2144 Intron NP_001269932.1
NM_001283004.1 2144 Intron NP_001269933.1
NM_001283005.1 2144 Intron NP_001269934.1
NM_001283006.1 2144 Intron NP_001269935.1
NM_001283007.1 2144 Intron NP_001269936.1
NM_001283008.1 2144 Intron NP_001269937.1
NM_003434.5 2144 Intron NP_003425.2
XM_005260819.1 2144 Intron XP_005260876.1
XM_005260820.4 2144 Intron XP_005260877.1
XM_011529336.1 2144 Intron XP_011527638.1
XM_011529337.1 2144 Intron XP_011527639.1
XM_011529338.2 2144 Intron XP_011527640.1
XM_011529339.1 2144 Intron XP_011527641.1
XM_011529340.1 2144 Intron XP_011527642.1
XM_017028041.1 2144 UTR 5 XP_016883530.1
XM_017028042.1 2144 Intron XP_016883531.1
XM_017028043.1 2144 UTR 5 XP_016883532.1
XM_017028044.1 2144 UTR 5 XP_016883533.1
XM_017028045.1 2144 UTR 5 XP_016883534.1
XM_017028046.1 2144 UTR 5 XP_016883535.1
XM_017028047.1 2144 Intron XP_016883536.1
XM_017028048.1 2144 Intron XP_016883537.1
XM_017028049.1 2144 UTR 5 XP_016883538.1
XM_017028050.1 2144 Intron XP_016883539.1
XM_017028051.1 2144 Intron XP_016883540.1
XM_017028052.1 2144 Intron XP_016883541.1
XM_017028053.1 2144 Intron XP_016883542.1
XM_017028054.1 2144 Intron XP_016883543.1
XM_017028055.1 2144 Intron XP_016883544.1
XM_017028056.1 2144 Intron XP_016883545.1
XM_017028057.1 2144 Intron XP_016883546.1
XM_017028058.1 2144 Intron XP_016883547.1

View Full Product Details