Product Details

SNP ID
rs117556279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18292210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCTATGTCCCGCTAAGGTACAT[A/G]AATCAGTGGAGCAAATAGAAGGAGA
Phenotype
MIM: 604075
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF133 PubMed Links

Gene Details

Gene
ZNF133
Gene Name
zinc finger protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083330.2 2393 Intron NP_001076799.1
NM_001282995.1 2393 Intron NP_001269924.1
NM_001282996.1 2393 Intron NP_001269925.1
NM_001282997.1 2393 Intron NP_001269926.1
NM_001282998.1 2393 Intron NP_001269927.1
NM_001282999.1 2393 Intron NP_001269928.1
NM_001283000.1 2393 Intron NP_001269929.1
NM_001283001.1 2393 Intron NP_001269930.1
NM_001283002.1 2393 Intron NP_001269931.1
NM_001283003.1 2393 Intron NP_001269932.1
NM_001283004.1 2393 Intron NP_001269933.1
NM_001283005.1 2393 Intron NP_001269934.1
NM_001283006.1 2393 Intron NP_001269935.1
NM_001283007.1 2393 Intron NP_001269936.1
NM_001283008.1 2393 Intron NP_001269937.1
NM_003434.5 2393 Intron NP_003425.2
XM_005260819.1 2393 Intron XP_005260876.1
XM_005260820.4 2393 Intron XP_005260877.1
XM_011529336.1 2393 Intron XP_011527638.1
XM_011529337.1 2393 Intron XP_011527639.1
XM_011529338.2 2393 Intron XP_011527640.1
XM_011529339.1 2393 Intron XP_011527641.1
XM_011529340.1 2393 Intron XP_011527642.1
XM_017028041.1 2393 UTR 5 XP_016883530.1
XM_017028042.1 2393 Intron XP_016883531.1
XM_017028043.1 2393 UTR 5 XP_016883532.1
XM_017028044.1 2393 UTR 5 XP_016883533.1
XM_017028045.1 2393 UTR 5 XP_016883534.1
XM_017028046.1 2393 UTR 5 XP_016883535.1
XM_017028047.1 2393 Intron XP_016883536.1
XM_017028048.1 2393 Intron XP_016883537.1
XM_017028049.1 2393 UTR 5 XP_016883538.1
XM_017028050.1 2393 Intron XP_016883539.1
XM_017028051.1 2393 Intron XP_016883540.1
XM_017028052.1 2393 Intron XP_016883541.1
XM_017028053.1 2393 Intron XP_016883542.1
XM_017028054.1 2393 Intron XP_016883543.1
XM_017028055.1 2393 Intron XP_016883544.1
XM_017028056.1 2393 Intron XP_016883545.1
XM_017028057.1 2393 Intron XP_016883546.1
XM_017028058.1 2393 Intron XP_016883547.1

View Full Product Details