Product Details

SNP ID
rs117240703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18297030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGTTGGTGGTTATGAGTCAACT[C/T]TTCCCTCATATCCCATAGGCCCTTT
Phenotype
MIM: 604075
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF133 PubMed Links

Gene Details

Gene
ZNF133
Gene Name
zinc finger protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083330.2 7213 Intron NP_001076799.1
NM_001282995.1 7213 Intron NP_001269924.1
NM_001282996.1 7213 Intron NP_001269925.1
NM_001282997.1 7213 Intron NP_001269926.1
NM_001282998.1 7213 Intron NP_001269927.1
NM_001282999.1 7213 Intron NP_001269928.1
NM_001283000.1 7213 Intron NP_001269929.1
NM_001283001.1 7213 Intron NP_001269930.1
NM_001283002.1 7213 Intron NP_001269931.1
NM_001283003.1 7213 Intron NP_001269932.1
NM_001283004.1 7213 Intron NP_001269933.1
NM_001283005.1 7213 Intron NP_001269934.1
NM_001283006.1 7213 Intron NP_001269935.1
NM_001283007.1 7213 Intron NP_001269936.1
NM_001283008.1 7213 Intron NP_001269937.1
NM_003434.5 7213 Intron NP_003425.2
XM_005260819.1 7213 Intron XP_005260876.1
XM_005260820.4 7213 Intron XP_005260877.1
XM_011529336.1 7213 Intron XP_011527638.1
XM_011529337.1 7213 Intron XP_011527639.1
XM_011529338.2 7213 Intron XP_011527640.1
XM_011529339.1 7213 Intron XP_011527641.1
XM_011529340.1 7213 Intron XP_011527642.1
XM_017028041.1 7213 UTR 5 XP_016883530.1
XM_017028042.1 7213 Intron XP_016883531.1
XM_017028043.1 7213 UTR 5 XP_016883532.1
XM_017028044.1 7213 UTR 5 XP_016883533.1
XM_017028045.1 7213 UTR 5 XP_016883534.1
XM_017028046.1 7213 UTR 5 XP_016883535.1
XM_017028047.1 7213 Intron XP_016883536.1
XM_017028048.1 7213 Intron XP_016883537.1
XM_017028049.1 7213 UTR 5 XP_016883538.1
XM_017028050.1 7213 Intron XP_016883539.1
XM_017028051.1 7213 Intron XP_016883540.1
XM_017028052.1 7213 Intron XP_016883541.1
XM_017028053.1 7213 Intron XP_016883542.1
XM_017028054.1 7213 Intron XP_016883543.1
XM_017028055.1 7213 Intron XP_016883544.1
XM_017028056.1 7213 Intron XP_016883545.1
XM_017028057.1 7213 Intron XP_016883546.1
XM_017028058.1 7213 Intron XP_016883547.1

View Full Product Details