Product Details
- SNP ID
-
rs117715076
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:21511485 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATTCTCTGTATTTTTAAAGTGTT[C/G]TCCACTTGCTTTAGAAGACGGCTGA
- Phenotype
-
MIM: 604612
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NKX2-2
PubMed Links
Gene Details
- Gene
- NKX2-2
- Gene Name
- NK2 homeobox 2
View Full Product Details