Product Details

SNP ID
rs118032296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2659105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGCAGATCAGAGGCAGGATGTGG[A/C]TACCTTCCTTGGAAGAAATAGAGAC
Phenotype
MIM: 604526 MIM: 614155 MIM: 614154
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IDH3B PubMed Links

Gene Details

Gene
IDH3B
Gene Name
isocitrate dehydrogenase 3 (NAD(+)) beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258384.2 1314 UTR 3 NP_001245313.1
NM_006899.4 1314 Intron NP_008830.2
NM_174855.3 1314 Intron NP_777280.1
XM_005260716.1 1314 Intron XP_005260773.1
Gene
MIR1292
Gene Name
microRNA 1292
There are no transcripts associated with this gene.

Gene
NOP56
Gene Name
NOP56 ribonucleoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006392.3 1314 Intron NP_006383.2
Gene
SNORA51
Gene Name
small nucleolar RNA, H/ACA box 51
There are no transcripts associated with this gene.

Gene
SNORD110
Gene Name
small nucleolar RNA, C/D box 110
There are no transcripts associated with this gene.

Gene
SNORD56
Gene Name
small nucleolar RNA, C/D box 56
There are no transcripts associated with this gene.

Gene
SNORD57
Gene Name
small nucleolar RNA, C/D box 57
There are no transcripts associated with this gene.

Gene
SNORD86
Gene Name
small nucleolar RNA, C/D box 86
There are no transcripts associated with this gene.

View Full Product Details