Product Details

SNP ID
rs116971075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30616477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATGATTTACTACAGCAACTATT[A/T]TGGTGGCTATGGGTATGGTGGGCTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP20-1 PubMed Links

Gene Details

Gene
KRTAP20-1
Gene Name
keratin associated protein 20-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181615.1 23 Missense Mutation TAT,TTT Y8F NP_853646.1
Gene
KRTAP20-4
Gene Name
keratin associated protein 20-4
There are no transcripts associated with this gene.

Gene
KRTAP6-1
Gene Name
keratin associated protein 6-1
There are no transcripts associated with this gene.

View Full Product Details