Product Details

SNP ID
rs116799330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30643005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACTGTGGCTACAGATGTGGCTG[G/T]GGTGGGGTATGGATTTGACTGCTGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP20-2 PubMed Links

Gene Details

Gene
KRTAP20-2
Gene Name
keratin associated protein 20-2
There are no transcripts associated with this gene.

Gene
KRTAP20-3
Gene Name
keratin associated protein 20-3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128077.1 142 Missense Mutation TGG,TGT W39C NP_001121549.1

View Full Product Details