Product Details

SNP ID
rs117596979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:16073067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTATGATTTCTTGGAATTGGGA[A/G]CACTTGACAAACTGAAGTAGAAAAA
Phenotype
MIM: 615964
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR99AHG PubMed Links

Gene Details

Gene
MIR99AHG
Gene Name
mir-99a-let-7c cluster host gene
There are no transcripts associated with this gene.

View Full Product Details