Product Details

SNP ID
rs114632703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:43791359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGTTTTGATGCAGGTGGTTTTA[C/T]GCACGACGAGCTGCTGAAGGTGTGG
Phenotype
MIM: 610653
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RRP1 PubMed Links

Gene Details

Gene
RRP1
Gene Name
ribosomal RNA processing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003683.5 227 Missense Mutation ACG,ATG T48M NP_003674.1
XM_017028485.1 227 Missense Mutation ACG,ATG T48M XP_016883974.1
XM_017028486.1 227 UTR 5 XP_016883975.1

View Full Product Details