Product Details

SNP ID
rs117709828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:45340829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTCCTTCTTTTCAGAAAGTGCA[A/G]ACCAAAGCTAAGACTGGCTCACACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00316 PubMed Links

Gene Details

Gene
LINC00316
Gene Name
long intergenic non-protein coding RNA 316
There are no transcripts associated with this gene.

View Full Product Details