Product Details

SNP ID
rs116322575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21919731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTTGAGCTACAGAGGCAGAATC[A/G]AACCAAAAACACTGCTTCCTTTAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPM1F PubMed Links

Gene Details

Gene
PPM1F
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014634.3 4873 UTR 3 NP_055449.1
XM_011530556.1 4873 UTR 3 XP_011528858.1
XM_017029108.1 4873 Intron XP_016884597.1

View Full Product Details