Product Details

SNP ID
rs115694446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39313831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGACCACAGGGCTGTTCTCAGA[A/G]GGAAGGCAACAAGGAACGGTTCCGC
Phenotype
MIM: 604163 MIM: 611069 MIM: 611068 MIM: 611070 MIM: 611071
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL3 PubMed Links

Gene Details

Gene
RPL3
Gene Name
ribosomal protein L3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000967.3 Intron NP_000958.1
NM_001033853.1 Intron NP_001029025.1
Gene
SNORD139
Gene Name
small nucleolar RNA, C/D box 139
There are no transcripts associated with this gene.

Gene
SNORD43
Gene Name
small nucleolar RNA, C/D box 43
There are no transcripts associated with this gene.

Gene
SNORD83A
Gene Name
small nucleolar RNA, C/D box 83A
There are no transcripts associated with this gene.

Gene
SNORD83B
Gene Name
small nucleolar RNA, C/D box 83B
There are no transcripts associated with this gene.

View Full Product Details