Product Details

SNP ID
rs117576785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:44004957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAATGGATGATTGATTTAAAAG[G/T]TTCTCAGGGACAGTGCTGTGTACTC
Phenotype
MIM: 608121 MIM: 612058
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PARVB PubMed Links

Gene Details

Gene
PARVB
Gene Name
parvin beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003828.2 Intron NP_001003828.1
NM_001243385.1 Intron NP_001230314.1
NM_001243386.1 Intron NP_001230315.1
NM_013327.4 Intron NP_037459.2
XM_005261596.1 Intron XP_005261653.1
XM_017028792.1 Intron XP_016884281.1
Gene
SAMM50
Gene Name
SAMM50 sorting and assembly machinery component
There are no transcripts associated with this gene.

View Full Product Details