Product Details

SNP ID
rs117194661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24543900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGTGGCCCGGTTGTAGCCACG[C/T]AGCACGATGAAGTGGCCCTGGTAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCD1 PubMed Links

Gene Details

Gene
GUCD1
Gene Name
guanylyl cyclase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284251.1 848 Silent Mutation CTA,CTG L246L NP_001271180.1
NM_001284252.1 848 Silent Mutation CTA,CTG L246L NP_001271181.1
NM_001284253.1 848 Silent Mutation CTA,CTG L203L NP_001271182.1
NM_001284254.1 848 Silent Mutation CTA,CTG L190L NP_001271183.1
NM_001284255.1 848 Silent Mutation CTA,CTG L147L NP_001271184.1
NM_001284256.1 848 Intron NP_001271185.1
NM_001284257.1 848 Intron NP_001271186.1
NM_031444.3 848 Silent Mutation CTA,CTG L190L NP_113632.2
XM_005261761.1 848 Intron XP_005261818.1

View Full Product Details