Product Details

SNP ID
rs111761876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:77900648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGAATCATTGGAGGATATTATTA[G/T]ATTGTGTCCTAAACTCTTAAGCAAC
Phenotype
MIM: 300885 MIM: 300715
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COX7B PubMed Links

Gene Details

Gene
COX7B
Gene Name
cytochrome c oxidase subunit 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001866.2 Intron NP_001857.1
Gene
MAGT1
Gene Name
magnesium transporter 1
There are no transcripts associated with this gene.

View Full Product Details