Product Details

SNP ID
rs111897802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:19708346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCAGCTCTAGCTGTCTCAGCTC[C/T]TCCAGCCCCACAGGCAGGTCCCGGC
Phenotype
MIM: 426000
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM5D PubMed Links

Gene Details

Gene
KDM5D
Gene Name
lysine demethylase 5D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146705.1 3624 Silent Mutation GAA,GAG E1084E NP_001140177.1
NM_001146706.1 3624 Silent Mutation GAA,GAG E996E NP_001140178.1
NM_004653.4 3624 Silent Mutation GAA,GAG E1053E NP_004644.2
XM_005262560.2 3624 Silent Mutation GAA,GAG E1008E XP_005262617.1
XM_005262561.2 3624 Silent Mutation GAA,GAG E976E XP_005262618.1
XM_011531468.2 3624 Silent Mutation GAA,GAG E1053E XP_011529770.1

View Full Product Details