Product Details

SNP ID
rs112622561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:20448363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGGCTGTGTGTCTGTTCGACTC[C/T]CCTCGCCCCATTTTCACCCCACCCC
Phenotype
MIM: 165370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RHOB PubMed Links

Gene Details

Gene
RHOB
Gene Name
ras homolog family member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004040.3 1293 UTR 3 NP_004031.1

View Full Product Details