Product Details

SNP ID
rs114488141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:184090780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGGTGTGGACAGGGCACAGAC[A/G]AGGGCCTGGTCTGAGGGGGTCCTTT
Phenotype
MIM: 616983
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENPP6 PubMed Links

Gene Details

Gene
ENPP6
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153343.3 1862 UTR 3 NP_699174.1

View Full Product Details