Product Details

SNP ID
rs115818714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185452285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGGTAGTTTTCTGCATCCATC[A/G]TCAATGAGTAATGGAAGAAATAAGC
Phenotype
MIM: 609488
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C4orf47 PubMed Links

Gene Details

Gene
C4orf47
Gene Name
chromosome 4 open reading frame 47
There are no transcripts associated with this gene.

Gene
CCDC110
Gene Name
coiled-coil domain containing 110
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145411.1 Intron NP_001138883.1
NM_152775.3 Intron NP_689988.1
XM_005262891.3 Intron XP_005262948.1
XM_006714172.2 Intron XP_006714235.1
XM_011531827.2 Intron XP_011530129.1
XM_011531828.2 Intron XP_011530130.1
XM_011531829.2 Intron XP_011530131.1
XM_017007987.1 Intron XP_016863476.1
XM_017007988.1 Intron XP_016863477.1

View Full Product Details