Product Details

SNP ID
rs116687668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:87655342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAAATACAAAACAAAGTGTTTT[C/T]CAAAGCAACAGCTCTTGAAGTGTGA
Phenotype
MIM: 600980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DMP1 PubMed Links

Gene Details

Gene
DMP1
Gene Name
dentin matrix acidic phosphoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079911.2 Intron NP_001073380.1
NM_004407.3 Intron NP_004398.1
XM_011531705.2 Intron XP_011530007.1
XM_011531706.2 Intron XP_011530008.1

View Full Product Details